This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04970099_m1
See other PLA1A GE Assays ›
Gene Symbol:
PLA1A
Gene Name:
phospholipase A1 member A
Gene Aliases:
PS-PLA1, PSPLA1
Chromosome Location:
Chr.3: 119597848 - 119629811 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04970099_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLA1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017006572.1 XP_016862061.1 2 - 3 163 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017006572.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phosphatidylserine metabolic process
lipid catabolic process
phosphatidylcholine 1-acylhydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0