This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04970111_m1
See other GTF2E1 GE Assays ›
Gene Symbol:
GTF2E1
Gene Name:
general transcription factor IIE subunit 1
Gene Aliases:
FE, TF2E1, TFIIE-A
Chromosome Location:
Chr.3: 120742711 - 120783134 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04970111_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2E1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005513.2 NP_005504.2 1 - 2 76 108
GenBank mRNA AK223401.1 - 1 - 2 42 108
BC005917.1 - 1 - 2 25 108
DA498425.1 - 1 - 2 76 108
S67859.1 - 1 - 2 25 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC005917.1, NM_005513.2, S67859.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF2E1

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
viral process
snRNA transcription from RNA polymerase II promoter
protein binding
sequence-specific DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000469772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000469772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000469772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000492959.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000492959.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000492959.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000492959.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000492959.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000283875.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000469772.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000469772.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000469772.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000484715.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000492959.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000492959.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000492959.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000492959.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000492959.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 ENST00000497393.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Clariom D Assay TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Clariom D Assay TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Clariom D Assay TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Clariom D Assay TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Clariom D Assay TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Clariom D Assay TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Clariom D Assay TC0300008518.hg.1 NM_005513.2 Refseq
Affymetrix Human Clariom D Assay TC0300008518.hg.1 NM_005513.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0