This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04970992_m1
See other LIPH GE Assays ›
Gene Symbol:
LIPH
Gene Name:
lipase H
Gene Aliases:
AH, ARWH2, HYPT7, LAH2, LPDLR, PLA1B, mPA-PLA1
Chromosome Location:
Chr.3: 185506262 - 185552661 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04970992_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011512531.2 XP_011510833.1 1 - 2 90 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011512531.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
lipid digestion
phospholipase activity
heparin binding
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000452897.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000452897.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000452897.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000452897.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0