This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04971005_g1
See other CLCN2 GE Assays ›
Gene Symbol:
CLCN2
Gene Name:
chloride voltage-gated channel 2
Gene Aliases:
CIC-2, CLC2, ECA2, ECA3, EGI11, EGI3, EGMA, EJM6, EJM8, LKPAT, clC-2
Chromosome Location:
Chr.3: 184346185 - 184361651 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04971005_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLCN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001171087.2 NP_001164558.1 18 - 19 2266 87
NM_001171088.2 NP_001164559.1 17 - 18 2185 87
NM_001171089.2 NP_001164560.1 18 - 19 2317 87
NM_004366.5 NP_004357.3 18 - 19 2317 87
XM_006713489.1 XP_006713552.1 18 - 19 2269 87
XM_006713490.2 XP_006713553.1 12 - 13 1527 87
XM_011512401.1 XP_011510703.1 18 - 19 2269 87
GenBank mRNA AF026004.1 - 18 - 19 2145 87
AK298952.1 - 18 - 19 2269 87
AK302759.1 - 17 - 18 2137 87
BC021578.1 - 8 - 9 1012 87
BC072004.1 - 18 - 19 2217 6424015 87
BX089717.1 - 1 - 2 127 87
KU177981.1 - 5 - 6 717 87
KU177982.1 - 5 - 6 717 87
KU177983.1 - 5 - 6 711 87
KU177984.1 - 5 - 6 645 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CLCN2

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transmembrane transport
retina development in camera-type eye
cell differentiation involved in salivary gland development
chloride transmembrane transport
regulation of anion transmembrane transport
voltage-gated chloride channel activity
chloride ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline