This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04972822_g1
See other EIF3J GE Assays ›
Gene Symbol:
EIF3J
Gene Name:
eukaryotic translation initiation factor 3 subunit J
Gene Aliases:
EIF3S1, eIF3-alpha, eIF3-p35
Chromosome Location:
Chr.15: 44537068 - 44562803 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04972822_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3J
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284336.1 NP_001271265.1 3 366 152
NM_003758.3 NP_003749.2 5 513 152
GenBank mRNA AF090923.1 - 1 109 152
AK002006.1 - 5 433 152
AK023388.1 - 5 463 152
BC002719.2 - 5 446 3632801 152
U97670.1 - 5 446 152

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF090923.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF3J

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
cytoplasmic translation
translational initiation
regulation of translational initiation
translation initiation factor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000536248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000536248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000536248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000536248.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558227.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558227.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558227.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000261868.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000424492.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000535391.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000536248.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000536248.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000536248.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000536248.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558053.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558053.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558053.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558053.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558053.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558227.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558227.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558227.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 ENST00000558353.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284335.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_001284336.1 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq
Affymetrix Human Clariom D Assay TC1500007093.hg.1 NM_003758.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0