This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04973088_g1
See other CDHR4 GE Assays ›
Gene Symbol:
CDHR4
Gene Name:
cadherin related family member 4
Gene Aliases:
CDH29, PRO34300
Chromosome Location:
Chr.3: 49790732 - 49802741 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04973088_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDHR4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017006366.1 XP_016861855.1 1 - 2 154 86
XM_017006368.1 XP_016861857.1 1 - 2 154 86
XM_017006369.1 XP_016861858.1 1 - 2 154 86
XM_017006370.1 XP_016861859.1 1 - 2 154 86
XM_017006371.1 XP_016861860.1 1 - 2 154 86

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017006366.1, XM_017006368.1, XM_017006369.1, XM_017006370.1, XM_017006371.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000462108.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000462108.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000462108.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000462108.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000462108.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000343366.8 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000412678.6 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000460430.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000462108.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000462108.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000462108.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000462108.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000462108.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487256.1 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 ENST00000487666.5 Ensembl
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq
Affymetrix Human Clariom D Assay TC0300011058.hg.1 NM_001007540.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9gdk9:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline