This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04973207_s1
See other LSAMP GE Assays ›
Gene Symbol:
LSAMP
Gene Name:
limbic system-associated membrane protein
Gene Aliases:
IGLON3, LAMP
Chromosome Location:
Chr.3: 115802363 - 116445542 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04973207_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LSAMP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318915.1 NP_001305844.1 9 7754 128
NM_002338.4 NP_002329.2 7 7685 128
GenBank mRNA BC041405.1 - 1 1019 128

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LSAMP

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
nervous system development
locomotory exploration behavior
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline