This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04973419_m1
See other IGSF10 GE Assays ›
Gene Symbol:
IGSF10
Gene Name:
immunoglobulin superfamily member 10
Gene Aliases:
CMF608
Chromosome Location:
Chr.3: 151435051 - 151619926 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04973419_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011512708.2 XP_011511010.1 2 - 3 227 97
XM_011512709.2 XP_011511011.1 2 - 3 227 97

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011512708.2, XM_011512709.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s)

ossification
multicellular organism development
cell differentiation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000489791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000489791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000497472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000497472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0