This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04973567_g1
See other PPP4R2 GE Assays ›
Gene Symbol:
PPP4R2
Gene Name:
protein phosphatase 4 regulatory subunit 2
Gene Aliases:
PP4R2
Chromosome Location:
Chr.3: 72996743 - 73069201 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04973567_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP4R2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AJ271448.2 - 1 401 143

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AJ271448.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300006444.hg.1 ENST00000423193.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006444.hg.1 ENST00000423193.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006444.hg.1 ENST00000423193.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006444.hg.1 ENST00000423193.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006444.hg.1 ENST00000423193.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006444.hg.1 ENST00000423193.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0