This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04973606_m1
See other NSUN7 GE Assays ›
Gene Symbol:
NSUN7
Gene Name:
NOP2/Sun RNA methyltransferase family member 7
Gene Aliases:
-
Chromosome Location:
Chr.4: 40749874 - 40810601 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04973606_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NSUN7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024677.4 NP_078953.3 6 - 7 1322 128
XM_017008611.1 XP_016864100.1 6 - 7 1345 128
XM_017008612.1 XP_016864101.1 6 - 7 1345 128
XM_017008613.1 XP_016864102.1 6 - 7 1345 128
XM_017008614.1 XP_016864103.1 6 - 7 1345 128
XM_017008615.1 XP_016864104.1 6 - 7 1345 128
GenBank mRNA AK304048.1 - 6 - 7 1046 128
BC036568.2 - 6 - 7 1294 5272990 128
BC042401.1 - 6 - 7 1322 128

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NSUN7

Gene Ontology Categories:

Function(s) Process(es)

methylation
RNA binding
methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000478857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000478857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000478857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 ENST00000478857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000316607.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000381782.6 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000423784.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000463952.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000469033.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000473399.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000478857.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000478857.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000478857.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 ENST00000478857.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq
Affymetrix Human Clariom D Assay TC0400007330.hg.1 NM_024677.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0