This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04974496_m1
See other FREM3 GE Assays ›
Gene Symbol:
FREM3
Gene Name:
FRAS1 related extracellular matrix 3
Gene Aliases:
-
Chromosome Location:
Chr.4: 143577407 - 143700675 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04974496_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FREM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AI184955.1 - 1 - 2 135 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000508899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000508899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000508899.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000508899.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0