This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04974603_m1
See other TRAPPC11 GE Assays ›
Gene Symbol:
TRAPPC11
Gene Name:
trafficking protein particle complex 11
Gene Aliases:
C4orf41, FOIGR, GRY, LGMD2S
Chromosome Location:
Chr.4: 183659267 - 183713594 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04974603_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021942.5 NP_068761.4 7 - 8 932 83
NM_199053.2 NP_951008.1 7 - 8 932 83
XM_017008537.1 XP_016864026.1 7 - 8 932 83
XM_017008538.1 XP_016864027.1 7 - 8 932 83
XM_017008539.1 XP_016864028.1 6 - 7 677 83
GenBank mRNA AK091146.1 - 2 - 3 156 83
AK096345.1 - 7 - 8 906 83
AK315057.1 - 2 - 3 138 83
AL833451.1 - 6 - 7 740 83
AL833571.1 - 7 - 8 911 83
BC051724.1 - 7 - 8 876 5265781 83
BX538044.1 - 7 - 8 886 83
BX647127.1 - 7 - 8 903 83
BX648499.1 - 7 - 8 907 83

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK091146.1, AK315057.1, XM_017008539.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRAPPC11

Gene Ontology Categories:

Function(s)

vesicle-mediated transport

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0