This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04974868_m1
See other PPARGC1A GE Assays ›
Gene Symbol:
PPARGC1A
Gene Name:
PPARG coactivator 1 alpha
Gene Aliases:
LEM6, PGC-1(alpha), PGC-1alpha, PGC-1v, PGC1, PGC1A, PPARGC1
Chromosome Location:
Chr.4: 23792021 - 24472975 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04974868_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPARGC1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011513768.1 XP_011512070.1 1 - 2 174 94
GenBank mRNA AK301883.1 - 1 - 2 153 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK301883.1, XM_011513768.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

response to reactive oxygen species
mitophagy
temperature homeostasis
cellular glucose homeostasis
negative regulation of protein phosphorylation
response to dietary excess
response to ischemia
galactose metabolic process
gluconeogenesis
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
mRNA processing
protein complex assembly
mitochondrion organization
positive regulation of transcription from RNA polymerase II promoter by pheromones
aging
digestion
circadian rhythm
androgen metabolic process
RNA splicing
response to cold
positive regulation of mitochondrion organization
skeletal muscle atrophy
response to muscle activity
response to electrical stimulus involved in regulation of muscle adaptation
negative regulation of smooth muscle cell migration
fatty acid oxidation
cerebellum development
respiratory electron transport chain
androgen receptor signaling pathway
forebrain development
circadian regulation of gene expression
cellular response to oxidative stress
positive regulation of histone acetylation
cellular response to potassium ion
response to statin
response to drug
response to starvation
regulation of circadian rhythm
response to leucine
negative regulation of neuron apoptotic process
cellular respiration
positive regulation of gluconeogenesis
negative regulation of glycolytic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fatty acid oxidation
positive regulation of smooth muscle cell proliferation
negative regulation of smooth muscle cell proliferation
protein stabilization
brown fat cell differentiation
positive regulation of sequence-specific DNA binding transcription factor activity
flavone metabolic process
adipose tissue development
cellular response to lipopolysaccharide
cellular response to nitrite
cellular response to caffeine
cellular response to fructose stimulus
cellular response to glucose stimulus
cellular response to interleukin-6
cellular response to tumor necrosis factor
cellular response to follicle-stimulating hormone stimulus
cellular response to estradiol stimulus
cellular response to hypoxia
cellular response to transforming growth factor beta stimulus
response to epinephrine
response to norepinephrine
negative regulation of mitochondrial fission
cellular response to thyroid hormone stimulus
negative regulation of neuron death
response to metformin
positive regulation of cellular respiration
positive regulation of mitochondrial DNA metabolic process
positive regulation of muscle tissue development
positive regulation of glomerular visceral epithelial cell apoptotic process
cellular response to ionomycin
cellular response to resveratrol
response to methionine
adaptive thermogenesis
positive regulation of progesterone biosynthetic process
negative regulation of receptor activity
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of energy homeostasis
positive regulation of ATP biosynthetic process
nucleotide binding
RNA polymerase II transcription cofactor activity
DNA binding
transcription coactivator activity
RNA binding
protein binding
transcription factor binding
ligand-dependent nuclear receptor binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
chromatin DNA binding
ubiquitin protein ligase binding
peroxisome proliferator activated receptor binding
alpha-tubulin binding
sequence-specific DNA binding
androgen receptor binding
promoter-specific chromatin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-7wltc:80/100.66.72.81:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline