This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04975023_m1
See other UGT8 GE Assays ›
Gene Symbol:
UGT8
Gene Name:
UDP glycosyltransferase 8
Gene Aliases:
CGT, UGT4
Chromosome Location:
Chr.4: 114597789 - 114687914 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04975023_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UGT8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322112.1 NP_001309041.1 2 - 3 676 159
GenBank mRNA BP219039.1 - 2 - 3 308 159

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001322112.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein localization to paranode region of axon
galactosylceramide biosynthetic process
glycosphingolipid metabolic process
cytoskeleton organization
central nervous system development
peripheral nervous system development
flavonoid biosynthetic process
paranodal junction assembly
neuron projection morphogenesis
flavonoid glucuronidation
2-hydroxyacylsphingosine 1-beta-galactosyltransferase activity
UDP-galactose:glucosylceramide beta-1,4-galactosyltransferase activity
glucuronosyltransferase activity
N-acylsphingosine galactosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0