This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04975041_m1
See other NDST3 GE Assays ›
Gene Symbol:
NDST3
Gene Name:
N-deacetylase and N-sulfotransferase 3
Gene Aliases:
HSST3
Chromosome Location:
Chr.4: 118033596 - 118258945 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04975041_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDST3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006714416.3 XP_006714479.1 1 - 2 305 96
XM_017008840.1 XP_016864329.1 1 - 2 87 96
GenBank mRNA AY358852.1 - 1 - 2 306 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY358852.1, XM_006714416.3, XM_017008840.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

heparan sulfate proteoglycan biosynthetic process
heparin biosynthetic process
[heparan sulfate]-glucosamine N-sulfotransferase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000394488.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000394488.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000394488.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000394488.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 ENST00000394488.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000296499.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000394488.2 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000394488.2 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000394488.2 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000394488.2 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 ENST00000394488.2 Ensembl
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq
Affymetrix Human Clariom D Assay TC0400008534.hg.1 NM_004784.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0