This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04975107_m1
See other SEC24D GE Assays ›
Gene Symbol:
SEC24D
Gene Name:
SEC24 homolog D, COPII coat complex component
Gene Aliases:
CLCRP2
Chromosome Location:
Chr.4: 118722823 - 118838683 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04975107_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEC24D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318066.1 NP_001304995.1 18 - 19 2677 150
NM_014822.3 NP_055637.2 18 - 19 2674 150
XM_005263379.2 XP_005263436.1 18 - 19 2551 150
XM_017008875.1 XP_016864364.1 13 - 14 1832 150
GenBank mRNA AB018298.1 - 18 - 19 2577 150
AF130464.2 - 17 - 18 2377 150
AK001390.1 - 15 - 16 2045 150
AK125962.1 - 12 - 13 1665 150
AK291765.1 - 18 - 19 2574 150
AK309960.1 - 11 - 12 1413 150
BC035761.1 - 18 - 19 2599 5554774 150
BC037736.1 - 21 - 22 3151 150

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SEC24D

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
antigen processing and presentation of peptide antigen via MHC class I
intracellular protein transport
ER to Golgi vesicle-mediated transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
COPII vesicle coating
molecular_function
protein binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0