This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04975323_s1
See other ADGRL3 GE Assays ›
Gene Symbol:
ADGRL3
Gene Name:
adhesion G protein-coupled receptor L3
Gene Aliases:
CIRL3, CL3, LEC3, LPHN3
Chromosome Location:
Chr.4: 61201256 - 62078335 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04975323_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322246.1 NP_001309175.1 26 10590 103
NM_001322402.1 NP_001309331.1 26 10742 103
NM_015236.5 NP_056051.2 25 10538 103
XM_011531788.2 XP_011530090.1 27 10298 103
XM_011531791.2 XP_011530093.1 24 10214 103
XM_017007929.1 XP_016863418.1 29 13200 103
XM_017007930.1 XP_016863419.1 28 12152 103
XM_017007931.1 XP_016863420.1 27 10409 103
XM_017007932.1 XP_016863421.1 26 10280 103
XM_017007933.1 XP_016863422.1 26 10271 103
XM_017007934.1 XP_016863423.1 26 10265 103
XM_017007936.1 XP_016863425.1 25 10241 103
XM_017007937.1 XP_016863426.1 27 11948 103
XM_017007939.1 XP_016863428.1 22 9995 103
XM_017007940.1 XP_016863429.1 27 10332 103
XM_017007941.1 XP_016863430.1 26 10305 103
XM_017007942.1 XP_016863431.1 26 10113 103
GenBank mRNA AK094703.1 - 1 126 103

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRL3

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
synapse assembly
locomotion involved in locomotory behavior
response to cocaine
positive regulation of synapse assembly
cell-cell adhesion
G-protein coupled receptor activity
calcium ion binding
protein binding
carbohydrate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0