This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04977333_m1
See other NNT GE Assays ›
Gene Symbol:
NNT
Gene Name:
nicotinamide nucleotide transhydrogenase
Gene Aliases:
GCCD4
Chromosome Location:
Chr.5: 43602574 - 43705566 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04977333_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NNT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005248275.4 XP_005248332.1 4 - 5 674 106

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005248275.4 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
NADPH regeneration
proton transport
cell redox homeostasis
oxidation-reduction process
reactive oxygen species metabolic process
hydrogen ion transmembrane transport
NAD(P)+ transhydrogenase (B-specific) activity
NAD(P)+ transhydrogenase activity
NAD(P)+ transhydrogenase (AB-specific) activity
NADP binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline