This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04977388_mH
See other MARVELD2 GE Assays ›
Gene Symbol:
MARVELD2
Gene Name:
MARVEL domain containing 2
Gene Aliases:
DFNB49, MARVD2, MRVLDC2, Tric
Chromosome Location:
Chr.5: 69415112 - 69444022 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04977388_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MARVELD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005248446.4 XP_005248503.1 1 - 2 147 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005248446.4 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

sensory perception of sound
cell-cell junction organization
establishment of endothelial barrier
bicellular tight junction assembly
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000515844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 ENST00000515844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000325631.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000413223.2 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000436532.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000454295.6 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000512803.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000515844.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 ENST00000515844.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001038603.2 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq
Affymetrix Human Clariom D Assay TC0500007676.hg.1 NM_001244734.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0