This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04978671_m1
See other STARD4 GE Assays ›
Gene Symbol:
STARD4
Gene Name:
StAR related lipid transfer domain containing 4
Gene Aliases:
-
Chromosome Location:
Chr.5: 111496033 - 111512597 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04978671_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308060.1 NP_001294989.1 1 - 2 207 99
GenBank mRNA AK315863.1 - 1 - 2 145 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK315863.1, NM_001308060.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
positive regulation of cholesterol esterification
positive regulation of bile acid biosynthetic process
cholesterol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline