This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04980365_g1
See other TMPRSS4-AS1 GE Assays ›
Gene Symbol:
TMPRSS4-AS1
Gene Name:
TMPRSS4 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 118015772 - 118086793 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04980365_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMPRSS4-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB231754.1 - 2 - 3 113 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000527695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 NR_038318.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 NR_038318.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 NR_038318.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012463.hg.1 NR_038318.1 Refseq
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527329.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527695.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527695.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000527695.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 ENST00000606951.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012463.hg.1 NR_038318.1 Refseq
Affymetrix Human Clariom D Assay TC1100012463.hg.1 NR_038318.1 Refseq
Affymetrix Human Clariom D Assay TC1100012463.hg.1 NR_038318.1 Refseq
Affymetrix Human Clariom D Assay TC1100012463.hg.1 NR_038318.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline