This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04980414_m1
See other IGSF9B GE Assays ›
Gene Symbol:
IGSF9B
Gene Name:
immunoglobulin superfamily member 9B
Gene Aliases:
-
Chromosome Location:
Chr.11: 133908564 - 133956960 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04980414_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF9B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277285.1 NP_001264214.1 19 - 20 4110 67
XM_006718795.3 XP_006718858.1 8 - 9 2624 67
XM_011542691.2 XP_011540993.1 20 - 21 4797 67
XM_011542692.2 XP_011540994.1 20 - 21 4785 67
XM_011542695.2 XP_011540997.1 19 - 20 4328 67
XM_017017393.1 XP_016872882.1 19 - 20 4815 67
XM_017017396.1 XP_016872885.1 13 - 14 4139 67
GenBank mRNA BG009128.1 - 1 - 2 238 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
nervous system development
positive regulation of inhibitory postsynaptic potential
kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000526663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000526663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000526663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000530957.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000530957.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000530957.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000321016.12 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000526663.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000526663.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000526663.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000527648.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000530957.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000530957.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000530957.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533160.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000533871.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 ENST00000564347.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012906.hg.1 NM_001277285.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0