This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04980509_s1
See other OPCML GE Assays ›
Gene Symbol:
OPCML
Gene Name:
opioid binding protein/cell adhesion molecule like
Gene Aliases:
IGLON1, OBCAM, OPCM
Chromosome Location:
Chr.11: 132403361 - 133532983 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04980509_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OPCML
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006718846.2 XP_006718909.1 8 18082 100
XM_011542856.2 XP_011541158.1 9 18395 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s)

cell adhesion
neuron recognition

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012881.hg.1 ENST00000624880.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012881.hg.1 ENST00000624880.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline