This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04981357_m1
See other ABLIM1 GE Assays ›
Gene Symbol:
ABLIM1
Gene Name:
actin binding LIM protein 1
Gene Aliases:
ABLIM, LIMAB1, LIMATIN, abLIM-1
Chromosome Location:
Chr.10: 114431110 - 114821643 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04981357_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABLIM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322884.1 NP_001309813.1 1 - 2 162 108
NM_001322885.1 NP_001309814.1 1 - 2 162 108
NM_001322886.1 NP_001309815.1 1 - 2 162 108
XM_017016245.1 XP_016871734.1 1 - 2 201 108
XM_017016250.1 XP_016871739.1 1 - 2 201 108
XM_017016255.1 XP_016871744.1 1 - 2 201 108
GenBank mRNA AK057834.1 - 1 - 2 197 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001322884.1, NM_001322885.1, NM_001322886.1, XM_017016245.1, XM_017016250.1, XM_017016255.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
visual perception
organ morphogenesis
lamellipodium assembly
cilium assembly
actin binding
protein binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700006942.hg.1 ENST00000458492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006942.hg.1 ENST00000458492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006942.hg.1 ENST00000458492.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006942.hg.1 ENST00000458492.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006942.hg.1 ENST00000458492.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006942.hg.1 ENST00000458492.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0