This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04981801_m1
See other GLUD1 GE Assays ›
Gene Symbol:
GLUD1
Gene Name:
glutamate dehydrogenase 1
Gene Aliases:
GDH, GDH1, GLUD
Chromosome Location:
Chr.10: 87050202 - 87095047 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04981801_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLUD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318906.1 NP_001305835.1 2 - 3 781 106
GenBank mRNA CD673630.1 - 2 - 3 266 106

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001318906.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glutamate biosynthetic process
glutamate catabolic process
glutamine metabolic process
cellular amino acid biosynthetic process
substantia nigra development
positive regulation of insulin secretion
oxidation-reduction process
tricarboxylic acid metabolic process
glutamate dehydrogenase (NAD+) activity
glutamate dehydrogenase [NAD(P)+] activity
protein binding
ATP binding
GTP binding
identical protein binding
ADP binding
NAD+ binding
leucine binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000399571.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000583339.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000583339.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000583339.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000399571.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000583339.3 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000583339.3 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000583339.3 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000630097.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 ENST00000631118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Clariom D Assay TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Clariom D Assay TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Clariom D Assay TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Clariom D Assay TC1000010536.hg.1 NR_111968.1 Refseq
Affymetrix Human Clariom D Assay TC1000010536.hg.1 NR_111968.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0