This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04982075_s1
See other GSTO2 GE Assays ›
Gene Symbol:
GSTO2
Gene Name:
glutathione S-transferase omega 2
Gene Aliases:
GSTO 2-2, bA127L20.1
Chromosome Location:
Chr.10: 104267626 - 104304948 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04982075_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTO2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011539270.2 XP_011537572.1 7 5762 126
XM_011539273.2 XP_011537575.1 5 4179 126
GenBank mRNA AK097894.1 - 1 420 126
AK126230.1 - 1 1326 126
AL831850.1 - 1 341 126

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
xenobiotic metabolic process
L-ascorbic acid metabolic process
oxidation-reduction process
cellular response to arsenic-containing substance
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
protein binding
oxidoreductase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0