This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04982510_s1
See other OAZ2 GE Assays ›
Gene Symbol:
OAZ2
Gene Name:
ornithine decarboxylase antizyme 2
Gene Aliases:
AZ2
Chromosome Location:
Chr.15: 64687574 - 64703281 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04982510_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OAZ2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301302.1 NP_001288231.1 5 1797 102
NM_002537.3 NP_002528.1 5 1800 102
GenBank mRNA AF057297.1 - 5 1719 102
BG059726.1 - 1 102
BM686393.1 - 1 92 102
BX648172.1 - 4 6954 102

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OAZ2

Panther Classification:

Molecular Function -

protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of cellular amino acid metabolic process
polyamine metabolic process
polyamine biosynthetic process
negative regulation of catalytic activity
positive regulation of protein catabolic process
positive regulation of intracellular protein transport
negative regulation of polyamine transmembrane transport
protein binding
ornithine decarboxylase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559555.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559555.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559665.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559665.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559665.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559665.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559912.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559912.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559912.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559912.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000559912.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560781.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560781.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560781.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560781.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000326005.10 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000558194.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559555.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559555.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559665.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559665.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559665.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559665.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559753.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559912.2 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559912.2 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559912.2 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559912.2 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000559912.2 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560258.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560781.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560781.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560781.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560781.6 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 ENST00000560837.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_001301302.1 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq
Affymetrix Human Clariom D Assay TC1500009753.hg.1 NM_002537.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0