This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04982950_m1
See other RBFOX1 GE Assays ›
Gene Symbol:
RBFOX1
Gene Name:
RNA binding protein, fox-1 homolog 1
Gene Aliases:
2BP1, A2BP1, FOX-1, FOX1, HRNBP1
Chromosome Location:
Chr.16: 5239749 - 7713343 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04982950_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBFOX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017023318.1 XP_016878807.1 392 98
XM_017023319.1 XP_016878808.1 2 - 3 388 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
RNA transport
nucleotide binding
RNA binding
mRNA binding
protein binding
protein C-terminus binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000526533.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000526533.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000526533.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000526533.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 NR_126335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 NR_126335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 NR_126335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009847.hg.1 NR_126335.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011348.hg.1 ENST00000585867.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000526533.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000526533.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000526533.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000526533.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 ENST00000527970.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009847.hg.1 NR_126335.1 Refseq
Affymetrix Human Clariom D Assay TC1100009847.hg.1 NR_126335.1 Refseq
Affymetrix Human Clariom D Assay TC1100009847.hg.1 NR_126335.1 Refseq
Affymetrix Human Clariom D Assay TC1100009847.hg.1 NR_126335.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rz6mw:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0