This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04984249_m1
See other PTP4A1 GE Assays ›
Gene Symbol:
PTP4A1
Gene Name:
protein tyrosine phosphatase type IVA, member 1
Gene Aliases:
HH72, PRL-1, PRL1, PTP(CAAX1), PTPCAAX1
Chromosome Location:
Chr.6: 63528021 - 63583588 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04984249_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTP4A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011536111.1 XP_011534413.1 1 - 2 51 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011536111.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
multicellular organism development
positive regulation of cell migration
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000473334.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000473334.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000473334.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000473334.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000578299.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000578299.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000578299.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000370651.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000473334.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000473334.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000473334.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000473334.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000578299.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000578299.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000578299.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000626021.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 ENST00000627002.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq
Affymetrix Human Clariom D Assay TC0600014142.hg.1 NM_003463.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6479c7f679-jmhj8:80/100.66.74.142:80.
git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.50.0-Offline