This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04984636_mH
See other DSE GE Assays ›
Gene Symbol:
DSE
Gene Name:
dermatan sulfate epimerase
Gene Aliases:
DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2
Chromosome Location:
Chr.6: 116254152 - 116441261 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04984636_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DSE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322938.1 NP_001309867.1 1 - 2 145 103
GenBank mRNA AK293872.1 - 1 - 2 124 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK293872.1, NM_001322938.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

heparan sulfate proteoglycan biosynthetic process
chondroitin sulfate biosynthetic process
dermatan sulfate biosynthetic process
chondroitin-glucuronate 5-epimerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0