This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04986990_g1
See other FRS3 GE Assays ›
Gene Symbol:
FRS3
Gene Name:
fibroblast growth factor receptor substrate 3
Gene Aliases:
FRS2-beta, FRS2B, FRS2beta, SNT-2, SNT2
Chromosome Location:
Chr.6: 41770176 - 41779905 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04986990_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FRS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017010193.1 XP_016865682.1 2 - 3 1727 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017010193.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
signal transduction
fibroblast growth factor receptor signaling pathway
positive regulation of GTPase activity
Ras guanyl-nucleotide exchange factor activity
fibroblast growth factor receptor binding
insulin receptor binding
protein binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000422888.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000422888.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000422888.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000422888.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000422888.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000426290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000426290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000426290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000426290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000426290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000466420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000466420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000466420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000466420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 ENST00000466420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000259748.6 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000373018.7 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000422888.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000422888.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000422888.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000422888.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000422888.5 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000426290.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000426290.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000426290.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000426290.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000426290.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000466420.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000466420.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000466420.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000466420.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 ENST00000466420.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq
Affymetrix Human Clariom D Assay TC0600011802.hg.1 NM_006653.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline