This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04989191_m1
See other AIMP2 GE Assays ›
Gene Symbol:
AIMP2
Gene Name:
aminoacyl tRNA synthetase complex interacting multifunctional protein 2
Gene Aliases:
JTV-1, JTV1, P38
Chromosome Location:
Chr.7: 6009241 - 6023834 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04989191_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AIMP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005249847.3 XP_005249904.1 1 - 2 461 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA aminoacylation for protein translation
apoptotic process
negative regulation of cell proliferation
positive regulation of protein ubiquitination
Type II pneumocyte differentiation
positive regulation of neuron death
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0