This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04989888_m1
See other AKR1D1 GE Assays ›
Gene Symbol:
AKR1D1
Gene Name:
aldo-keto reductase family 1, member D1
Gene Aliases:
3o5bred, CBAS2, SRD5B1
Chromosome Location:
Chr.7: 138076432 - 138118304 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04989888_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AKR1D1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190906.1 NP_001177835.1 1 - 2 170 64
NM_001190907.1 NP_001177836.1 1 - 2 170 64
NM_005989.3 NP_005980.1 1 - 2 170 64
GenBank mRNA AK289425.1 - 1 - 2 110 64
AK298421.1 - 1 - 2 171 64
AK298428.1 - 1 - 2 136 64
BC130625.1 - 1 - 2 125 40146656 64
BC130627.1 - 1 - 2 125 40146658 64
FM160741.1 - 4 - 5 666 64
Z28339.1 - 1 - 2 152 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AKR1D1

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

bile acid biosynthetic process
cholesterol catabolic process
digestion
C21-steroid hormone metabolic process
androgen metabolic process
bile acid catabolic process
oxidation-reduction process
aldo-keto reductase (NADP) activity
steroid binding
delta4-3-oxosteroid 5beta-reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0