This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04990262_m1
See other BET1 GE Assays ›
Gene Symbol:
BET1
Gene Name:
Bet1 golgi vesicular membrane trafficking protein
Gene Aliases:
HBET1
Chromosome Location:
Chr.7: 93962770 - 94004382 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04990262_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BET1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_133908.1 5 - 6 592 126
GenBank mRNA CR749267.1 - 5 - 6 543 126

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
protein transport
COPII vesicle coating
vesicle fusion with Golgi apparatus
protein binding
syntaxin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000424151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000424151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000425626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000425626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000425626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000425626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000425626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000471446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000471446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000471446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 ENST00000471446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000222547.7 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000357520.8 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000424151.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000424151.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000425626.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000425626.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000425626.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000425626.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000425626.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000433727.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000457139.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000471446.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000471446.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000471446.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 ENST00000471446.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Clariom D Assay TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Clariom D Assay TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Clariom D Assay TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Clariom D Assay TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Clariom D Assay TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Clariom D Assay TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Clariom D Assay TC0700011813.hg.1 NM_005868.5 Refseq
Affymetrix Human Clariom D Assay TC0700011813.hg.1 NM_005868.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rw96b:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0