This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04992115_m1
See other GTF2IP23 GE Assays ›
Gene Symbol:
GTF2IP23
Gene Name:
general transcription factor IIi pseudogene 23
Gene Aliases:
-
Chromosome Location:
Chr.7: 66847450 - 66884273 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04992115_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2IP23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_135738.1 3 - 4 350 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000412091.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000412091.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000412091.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000412091.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000412091.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000424928.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000424928.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000441664.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000441664.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000412091.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000412091.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000412091.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000412091.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000412091.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000424928.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000424928.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000429320.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000441664.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000441664.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013571.hg.1 ENST00000450353.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline