This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04993317_m1
See other PRSS55 GE Assays ›
Gene Symbol:
PRSS55
Gene Name:
protease, serine 55
Gene Aliases:
CT153, T-SP1, TSP1, UNQ9391
Chromosome Location:
Chr.8: 10525331 - 10554166 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04993317_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS55
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC064412.1 - 2 - 3 705 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

serine protease

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000328655.7 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000518641.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 ENST00000522210.1 Ensembl
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_001197020.1 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_198464.3 Refseq
Affymetrix Human Clariom D Assay TC0800006715.hg.1 NM_198464.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-8rs9g:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline