This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04993338_m1
See other EXTL3 GE Assays ›
Gene Symbol:
EXTL3
Gene Name:
exostosin like glycosyltransferase 3
Gene Aliases:
BOTV, EXTL1L, EXTR1, REGR, RPR
Chromosome Location:
Chr.8: 28701473 - 28753690 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04993338_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EXTL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001440.3 NP_001431.1 2 - 3 288 72
NR_073469.1 2 - 3 288 72
GenBank mRNA AB007042.1 - 2 - 3 125 72
AB011091.1 - 2 - 3 147 72
AF001690.1 - 2 - 3 133 72
AF083551.1 - 2 - 3 125 72
BI911162.1 - 2 - 3 156 72
DA757362.1 - 2 - 3 438 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB007042.1, AB011091.1, AF001690.1, AF083551.1, NM_001440.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EXTL3

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
positive regulation of cell growth
IRE1-mediated unfolded protein response
glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
receptor activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0