This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04993635_gH
See other EEF1D GE Assays ›
Gene Symbol:
EEF1D
Gene Name:
eukaryotic translation elongation factor 1 delta
Gene Aliases:
EF-1D, EF1D, FP1047
Chromosome Location:
Chr.8: 143579697 - 143597675 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04993635_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130056.2 NP_001123528.1 2 - 3 454 97
NM_001317743.1 NP_001304672.1 2 - 3 451 97
XM_006716520.3 XP_006716583.1 3 - 4 6551 97
XM_017013172.1 XP_016868661.1 3 - 4 1539 97
XM_017013173.1 XP_016868662.1 2 - 3 631 97
GenBank mRNA BC000678.2 - 2 - 3 998 97
BC094806.1 - 2 - 3 165 97
BG482145.1 - 2 - 3 158 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
cellular response to ionizing radiation
cell-cell adhesion
DNA binding
translation elongation factor activity
signal transducer activity
protein binding
translation factor activity, RNA binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0