This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04994253_m1
See other EYA1 GE Assays ›
Gene Symbol:
EYA1
Gene Name:
EYA transcriptional coactivator and phosphatase 1
Gene Aliases:
BOP, BOR, BOS1, OFC1
Chromosome Location:
Chr.8: 71197433 - 71548130 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04994253_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EYA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017013206.1 XP_016868695.1 2 - 3 255 137
GenBank mRNA BC035138.1 - 2 - 3 259 137
BC048982.1 - 2 - 3 253 137

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017013206.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
metanephros development
branching involved in ureteric bud morphogenesis
outflow tract morphogenesis
double-strand break repair
transcription, DNA-templated
pattern specification process
mesodermal cell fate specification
sensory perception of sound
anatomical structure morphogenesis
response to ionizing radiation
striated muscle tissue development
histone dephosphorylation
protein sumoylation
cellular protein localization
establishment or maintenance of apical/basal cell polarity
peptidyl-tyrosine dephosphorylation
aorta morphogenesis
outer ear morphogenesis
middle ear morphogenesis
regulation of neuron differentiation
positive regulation of DNA repair
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
neuron fate specification
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
positive regulation of epithelial cell proliferation
pharyngeal system development
lung epithelial cell differentiation
otic vesicle morphogenesis
positive regulation of secondary heart field cardioblast proliferation
cochlea morphogenesis
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
RNA binding
protein tyrosine phosphatase activity
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000519927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000519927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000519927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000519927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000519927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000523987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000523987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000523987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012425.hg.1 ENST00000523987.4 Ensembl
Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000519927.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000519927.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000519927.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000519927.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000519927.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000523987.4 Ensembl
Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000523987.4 Ensembl
Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000523987.4 Ensembl
Affymetrix Human Clariom D Assay TC0800012425.hg.1 ENST00000523987.4 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0