This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04995171_s1
See other ARC GE Assays ›
Gene Symbol:
ARC
Gene Name:
activity regulated cytoskeleton associated protein
Gene Aliases:
Arg3.1
Chromosome Location:
Chr.8: 142611044 - 142614472 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04995171_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015193.4 NP_056008.1 1 527 77
GenBank mRNA AF193421.1 - 1 515 77
BC012321.1 - 1 531 4555433 77
BC075802.1 - 1 519 5198843 77
BI752734.1 - 1 521 77
D87468.1 - 1 519 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARC

Gene Ontology Categories:

Function(s)

endocytosis
cytoskeleton organization
endoderm development
learning
anterior/posterior pattern specification
cell migration
regulation of cell morphogenesis
regulation of neuronal synaptic plasticity
positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 ENST00000581404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 ENST00000581404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 ENST00000581404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Clariom D Assay TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Clariom D Assay TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Clariom D Assay TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Clariom D Assay TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Clariom D Assay TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Clariom D Assay TC0800012076.hg.1 ENST00000356613.3 Ensembl
Affymetrix Human Clariom D Assay TC0800012076.hg.1 ENST00000581404.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012076.hg.1 ENST00000581404.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012076.hg.1 ENST00000581404.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Clariom D Assay TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Clariom D Assay TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Clariom D Assay TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Clariom D Assay TC0800012076.hg.1 NM_015193.4 Refseq
Affymetrix Human Clariom D Assay TC0800012076.hg.1 NM_015193.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0