This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04996714_m1
See other ELAVL2 GE Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like neuron-specific RNA binding protein 2
Gene Aliases:
HEL-N1, HELN1, HUB
Chromosome Location:
Chr.9: 23690099 - 23850757 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04996714_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELAVL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006716734.3 XP_006716797.1 1 - 2 553 112
XM_011517774.2 XP_011516076.1 1 - 2 567 112
XM_011517777.2 XP_011516079.1 1 - 2 553 112
XM_017014411.1 XP_016869900.1 1 - 2 536 112
XM_017014414.1 XP_016869903.1 1 - 2 540 112

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006716734.3, XM_011517774.2, XM_011517777.2, XM_017014411.1, XM_017014414.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
regulation of transcription, DNA-templated
nucleotide binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000423281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000423281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000423281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000423281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000423281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000440102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000440102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000440102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000440102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000440102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000462649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000462649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000462649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000223951.10 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380110.8 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000380117.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000397312.6 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000423281.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000423281.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000423281.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000423281.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000423281.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000440102.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000440102.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000440102.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000440102.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000440102.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000462649.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000462649.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000462649.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 ENST00000544538.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171195.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_001171197.1 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq
Affymetrix Human Clariom D Assay TC0900009740.hg.1 NM_004432.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0