This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04998449_m1
See other NHS GE Assays ›
Gene Symbol:
NHS
Gene Name:
NHS actin remodeling regulator
Gene Aliases:
CTRCT40, CXN, SCML1
Chromosome Location:
Chr.X: 17375420 - 17735994 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04998449_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NHS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136024.3 NP_001129496.1 2 - 3 464 80
NM_001291867.1 NP_001278796.1 2 - 3 1059 80
NM_001291868.1 NP_001278797.1 2 - 3 464 80
NM_198270.3 NP_938011.1 2 - 3 1059 80
GenBank mRNA AK302437.1 - 2 - 3 1245 80
AY436752.1 - 2 - 3 1059 80
AY456992.1 - 2 - 3 464 80
AY456993.1 - 1 - 2 1119 80
BC136415.1 - 2 - 3 785 9020403 80
BC171763.1 - 2 - 3 785 9054416 80
CR936788.1 - 2 - 3 233 80
GQ988776.1 - 2 - 3 721 80

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NHS

Gene Ontology Categories:

Function(s)

lens development in camera-type eye
cell differentiation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0