This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04998450_m1
See other NHS GE Assays ›
Gene Symbol:
NHS
Gene Name:
NHS actin remodeling regulator
Gene Aliases:
CTRCT40, CXN, SCML1
Chromosome Location:
Chr.X: 17375420 - 17735994 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04998450_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NHS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136024.3 NP_001129496.1 3 - 4 597 87
NM_001291867.1 NP_001278796.1 3 - 4 1192 87
GenBank mRNA AK302437.1 - 3 - 4 1378 87
AY456992.1 - 3 - 4 597 87
AY456993.1 - 2 - 3 1252 87
BC136415.1 - 3 - 4 918 9020403 87
BC171763.1 - 3 - 4 918 9054416 87
GQ988776.1 - 3 - 4 854 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s)

lens development in camera-type eye
cell differentiation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000380060.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000398097.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000485305.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000615422.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 ENST00000617601.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001136024.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291867.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_001291868.1 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq
Affymetrix Human Clariom D Assay TC0X00006709.hg.1 NM_198270.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0