This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04998455_m1
See other NHS GE Assays ›
Gene Symbol:
NHS
Gene Name:
NHS actin remodeling regulator
Gene Aliases:
CTRCT40, CXN, SCML1
Chromosome Location:
Chr.X: 17375420 - 17735994 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04998455_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NHS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136024.3 NP_001129496.1 8 - 9 4085 92
NM_001291867.1 NP_001278796.1 8 - 9 4680 92
NM_001291868.1 NP_001278797.1 7 - 8 4022 92
NM_198270.3 NP_938011.1 7 - 8 4617 92
XM_011545528.2 XP_011543830.1 5 - 6 4229 92
GenBank mRNA AY436752.1 - 7 - 8 4617 92
AY456992.1 - 8 - 9 4085 92
AY456993.1 - 7 - 8 4740 92
BC136415.1 - 8 - 9 4406 9020403 92
BC171763.1 - 8 - 9 4406 9054416 92
CR749300.1 - 2 - 3 1455 92
CR936788.1 - 7 - 8 3791 92
GQ988776.1 - 8 - 9 4345 92

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NHS

Gene Ontology Categories:

Function(s)

lens development in camera-type eye
cell differentiation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bq2hq:80/100.66.76.182:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0