This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04998633_m1
See other AWAT1 GE Assays ›
Gene Symbol:
AWAT1
Gene Name:
acyl-CoA wax alcohol acyltransferase 1
Gene Aliases:
DGA2, DGAT2L3
Chromosome Location:
Chr.X: 70233489 - 70240661 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04998633_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AWAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013579.2 NP_001013597.1 3 - 4 301 120
XM_017029295.1 XP_016884784.1 3 - 4 1467 120
GenBank mRNA AY947638.1 - 3 - 4 260 120
W68831.1 - 3 - 4 301 120

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AWAT1

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

wax biosynthetic process
arachidonic acid metabolic process
long-chain-alcohol O-fatty-acyltransferase activity
wax ester synthase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000374521.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 ENST00000480702.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 NM_001013579.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007532.hg.1 NM_001013579.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0