This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04998730_m1
See other VSIG1 GE Assays ›
Gene Symbol:
VSIG1
Gene Name:
V-set and immunoglobulin domain containing 1
Gene Aliases:
1700062D20Rik, GPA34, dJ889N15.1
Chromosome Location:
Chr.X: 108018886 - 108079184 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04998730_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VSIG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170553.1 NP_001164024.1 2 - 3 376 87
XM_005262127.4 XP_005262184.1 2 - 3 400 87
XM_011530936.2 XP_011529238.1 2 - 3 259 87
GenBank mRNA AK301311.1 - 2 - 3 276 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s)

epithelial cell morphogenesis
maintenance of gastrointestinal epithelium

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000479635.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000479635.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000479635.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000479635.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000479635.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000479635.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay TC0X00008080.hg.1 NM_182607.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-c7pcq:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0