This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04998855_s1
See other PAK3 GE Assays ›
Gene Symbol:
PAK3
Gene Name:
p21 (RAC1) activated kinase 3
Gene Aliases:
ARA, MRX30, MRX47, OPHN3, PAK-3, PAK3beta, bPAK, beta-PAK
Chromosome Location:
Chr.X: 110944241 - 111227361 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04998855_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PAK3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_136740.1 3 5993 119
NR_136741.1 4 6060 119
NR_136742.1 5 6208 119
NR_136743.1 4 6144 119
NR_136744.1 5 6211 119
NR_136745.1 6 5239 119
NR_136746.1 6 6403 119
GenBank mRNA BX537613.1 - 1 4422 119

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPK activity
mitotic cell cycle
stimulatory C-type lectin receptor signaling pathway
axonogenesis
dendrite development
regulation of actin filament polymerization
T cell costimulation
positive regulation of neuron apoptotic process
ephrin receptor signaling pathway
synapse organization
T cell receptor signaling pathway
dendritic spine morphogenesis
positive regulation of dendritic spine morphogenesis
cellular response to organic cyclic compound
positive regulation of DNA biosynthetic process
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Rho GTPase binding
SH3 domain binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0