This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04998965_s1
See other GPRASP1 GE Assays ›
Gene Symbol:
GPRASP1
Gene Name:
G protein-coupled receptor associated sorting protein 1
Gene Aliases:
GASP, GASP-1, GASP1
Chromosome Location:
Chr.X: 102651366 - 102659083 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04998965_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPRASP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001099410.1 NP_001092880.1 4 3931 97
NM_001099411.1 NP_001092881.1 3 3863 97
NM_001184727.1 NP_001171656.1 6 4034 97
NM_014710.4 NP_055525.3 5 4022 97
XM_017029981.1 XP_016885470.1 4 3911 97
XM_017029982.1 XP_016885471.1 4 3946 97
GenBank mRNA AB007903.2 - 1 3387 97
BC114552.1 - 3 3667 40037190 97

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPRASP1

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

endosome to lysosome transport
G-protein coupled receptor catabolic process
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0