This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05001833_g1
See other PPA1 GE Assays ›
Gene Symbol:
PPA1
Gene Name:
pyrophosphatase (inorganic) 1
Gene Aliases:
HEL-S-66p, IOPPP, PP, PP1, SID6-8061
Chromosome Location:
Chr.10: 70202830 - 70233434 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05001833_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021129.3 NP_066952.1 2 - 3 227 76
GenBank mRNA AB026723.1 - 2 - 3 150 76
AF092439.1 - 2 - 3 197 76
AF108211.1 - 2 - 3 108 76
AF119665.1 - 2 - 3 197 76
AF154065.1 - 2 - 3 165 76
AF217186.1 - 2 - 3 120 76
AK058193.1 - 2 - 3 220 76
BC001022.2 - 2 - 3 202 3345496 76
BC061581.1 - 2 - 3 210 6526628 76
BC105034.1 - 2 - 3 193 8144037 76
BC105036.1 - 2 - 3 193 8144039 76
BC107882.1 - 2 - 3 161 6495107 76
BU520836.1 - 2 - 3 208 76
EU794626.1 - 2 - 3 227 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPA1

Panther Classification:

Molecular Function -

pyrophosphatase

Gene Ontology Categories:

Function(s) Process(es)

tRNA aminoacylation for protein translation
phosphate-containing compound metabolic process
diphosphate metabolic process
magnesium ion binding
inorganic diphosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000460755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000460755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000460755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000495346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000495346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000495346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009479.hg.1 ENST00000523057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010455.hg.1 ENST00000393779.2 Ensembl
Affymetrix Human Clariom D Assay TC0300010455.hg.1 ENST00000393779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009479.hg.1 ENST00000523057.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline