This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05001974_m1
See other GSTO2 GE Assays ›
Gene Symbol:
GSTO2
Gene Name:
glutathione S-transferase omega 2
Gene Aliases:
GSTO 2-2, bA127L20.1
Chromosome Location:
Chr.10: 104267626 - 104304948 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05001974_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTO2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011539271.1 XP_011537573.1 5 - 6 782 119
XM_011539272.2 XP_011537574.1 5 - 6 782 119
GenBank mRNA AK300508.1 - 4 - 5 490 119

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
xenobiotic metabolic process
L-ascorbic acid metabolic process
oxidation-reduction process
cellular response to arsenic-containing substance
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
protein binding
oxidoreductase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000467629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000467629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000467629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000467629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000467629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000473401.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000473401.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000473401.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000473401.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000473401.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000498052.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000498052.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000498052.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000498052.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007253.hg.1 ENST00000579240.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000467629.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000467629.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000467629.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000467629.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000467629.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000473401.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000473401.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000473401.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000473401.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000473401.5 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000498052.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000498052.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000498052.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000498052.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
Affymetrix Human Clariom D Assay TC1600007253.hg.1 ENST00000579240.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0